Would you like to log in again?
Whatman™ Anopore Inorganic Membranes Without Support Ring, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Anodisc™ Anodisc™
6809-7023 6809-5522 6809-7013 6809-5502 6809-7003 6809-7003 6809-7023 6809-7013 6809-5522 6809-5502
89203-116PK 900.6 USD InStock
CA89203-116 89203-116 CA28138-061 CA28138-057 28138-059 28138-057 CA28138-059 28138-061 11025-678 CA11025-678
Whatman™ Anopore Inorganic Membranes Without Support Ring, Whatman products (Cytiva)
Filters Membrane Filters
Whatman Anopore inorganic membranes, supplied as Anodisc membrane filters, are an aluminum oxide membrane with a honeycomb pore structure that facilitates highly precise filtration. Their transparency and surface retention enable efficient light or electron microscopy.
  • Virtually transparent when wet
  • Minimal autofluorescence
  • Hydrophilic with wide solvent compatibility
  • Non-toxic for cellular growth
The non-deformable, capillary pore structure has no lateral crossovers between individual pores. The inorganic membrane filters have high pore density and narrow pore size, allowing particles no larger than the specified size to pass through the filter.
Microorganisms and particulates are captured on the surface of the membrane for light microscopy or electron microscopy sample preparation so they can be examined without transferring to another surface. No monomers, plasticizers, adhesives, surfactants or wetting agents are used in the manufacturing process, which eliminates sample contamination and ensures low protein binding and minimal loss of sample.
The Anodisc membrane filters enable easier handling and are suitable for both vacuum and pressure filtration.
Whatman Anodisc membrane filters are available in a variety of pore sizes and membrane diameters and in format with and without support rings.
Anodisc membrane filters can also be used for liposome extrusion, HPLC mobile phase filtration and degassing, solvent cleaning, and gravimetric analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty