Would you like to log in again?
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ GD/X™
6869-2502 6872-1304 6880-1302 6875-1302 6905-2502 6878-1304 6872-2504 6894-1304 6878-1302 6880-2504 6888-2527 6882-2516 6880-2502 6882-1316 6875-2504 6871-2502 6878-2502 6895-2504 6888-1327 6870-2504 6879-2504 6874-2502 6876-2502 6890-1307 6874-1304 6876-1302 6891-2507 6871-2504 6871-1304 6892-2515 6884-1310 6871-2550 6869-2504 6881-2504 6888-1327 6891-2507 6875-2502 6888-2527 6872-2504 6871-1302 6871-1302 6869-2504 6876-1304 6880-1304 6874-2504 6894-1304 6870-2504 6892-2515 6874-1304 6878-1302 6872-1302 6882-2504 6894-2504 6888-2502 6887-2502 6880-1302 6886-2512 6871-2550 6874-1302 6888-2502 6905-2502 6878-2502 6869-2502 6872-1302 6894-2504 6872-2502 6873-2502 6883-2504 6870-2550 6876-2504 6880-2504 6875-2504 6870-2502 6882-2516 6874-1302 6905-2504 6884-2510 6876-2502 6886-1312 6870-2502 6872-1304 6882-2504 6886-2512 6876-2504 6878-1304 6890-2507 6874-2502 6871-1304 6890-2507 6870-1302 6872-2502 6883-2516 6873-1304 6882-1316 6886-1312 6878-2504 6870-1304 6879-2504 6875-1302 6905-2504 6875-1304 6884-2510 6870-1302 6870-1304 6875-2502 6880-1304 6874-2504 6871-2502 6884-1310 6878-2504 6871-2504 6883-2504 6876-1302 6875-1304 6883-2516 6876-1304 6873-1304 6873-2504 6870-2550 6890-1307 6881-2504 6895-2504 6887-2502 6880-2502
89233-778PK 4095.83 USD InStock
89233-778 CA80087-208 CA89233-784 CA89233-782 CA89233-780 28138-182 CA28138-192 28138-180 CA28138-190 28138-186 28138-184 CA28138-194 28138-102 CA28138-112 28138-188 CA28138-110 28138-106 28138-104 CA28138-114 14217-554 28138-108 CA28138-118 CA14223-314 CA89233-778 80087-208 CA28138-180 28138-172 28138-170 CA28138-184 28138-176 14005-864 CA28138-182 28138-174 CA28138-188 CA28138-186 28138-178 CA28138-104 CA28138-102 CA28138-108 14223-314 75831-558 CA28138-106 70240-004 28138-160 CA28138-170 28138-164 CA28138-174 28138-162 CA28138-172 28138-168 28138-202 CA28138-178 28138-166 CA28138-176 CA14005-864 28138-200 28138-204 28138-150 CA11013-180 CA28138-162 28138-154 CA28138-160 CA28138-166 CA28138-200 28138-158 CA28138-164 28138-156 CA28138-204 CA28138-168 CA28138-202 CA10147-962 CA11012-630 28138-142 28138-140 CA11012-628 CA28138-150 28138-146 CA28138-156 CA28138-154 CA11008-528 CA28138-158 CA70240-004 CA28138-140 CA11012-638 CA75830-324 CA28138-142 CA28138-146 28138-138 10147-970 10147-972 28138-120 28138-124 10147-968 CA28138-138 10147-966 28138-126 10147-960 10147-964 10035-516 CA10147-972 10035-518 10147-962 CA10147-970 89233-782 28138-190 89233-784 28138-194 89233-780 28138-192 28138-110 28138-198 28138-196 28138-114 CA14217-554 CA28138-120 28138-112 CA28138-126 CA10147-966 28138-118 CA10147-964 CA28138-124 10035-520 CA10147-968 10035-522
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Filters Syringe Filters
Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.
  • Recyclable after use
Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Cytiva also offers Sterile Whatman GD/X syringe filters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty