Would you like to log in again?
Whatman™ GD/X Sterile Syringe Filters
Supplier: Whatman products (Cytiva)
Whatman™
6901-2502 6896-2504 6901-2504 6900-2504 6900-2504 6902-2504 6896-2502 6901-2504 6900-2502 6901-2502 6900-2502 6896-2502 6896-2504 6902-2504
28138-400PK 268.85 USD InStock
CA28138-408 28138-400 28138-404 CA28138-402 28138-206 28138-402 CA28138-400 CA28138-208 28138-408 CA28138-406 28138-406 CA28138-206 CA28138-404 28138-208
Whatman™ GD/X Sterile Syringe Filters
Filters Syringe Filters
Cytiva's Whatman GD/X sterile syringe filter is designed to support filtration of viscous, hard-to-filter samples with high solids content.
  • Reduces need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters
  • Female luer lock for leak-free connections
  • High-efficiency Whatman GMF 150 and GF/F pre-filter media
  • Eliminates or greatly reduces the need to carry out separate prefiltration step for difficult-to-filter samples
  • Recyclable after use
Cytiva's GD/X sterile syringe filter is suitable for preparing viscous, dense, or otherwise hard-to-filter samples prior to a variety of analytical methods.
Sterile GD/X syringe filters combine a polypropylene housing, Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES)
- Polyvinylidene difluoride (PVDF)
- Cellulose acetate
- Glass microfiber
Cytiva also offers non sterile Whatman GD/X syringe filters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty