Would you like to log in again?
GWV High-Capacity Groundwater Sampling Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12025 12020 12179 12025 12180 12179 12180 12020
28145-148PK 1867.7 USD InStock
28145-148 CA28145-147 28100-052 28139-434 CA28139-434 CA28145-148 CA28100-052 28145-147
GWV High-Capacity Groundwater Sampling Capsules, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
These disposable filters are designed specifically for the filtration of groundwater prior to dissolved metals analysis.
  • Disposable
  • For groundwater
  • Polypropylene housing
They provide an easy, efficient way to meet filtration requirements of the EPA or other regulatory agency protocols and eliminate the costly decontamination, assembly/disassembly, and multiple filter membrane changes associated with reusable filter holders. Filter media is Versapor® membrane, an acrylic copolymer on a nonwoven support. Polypropylene housing. Inlet/outlet connections: 3.2 mm (¹/₈") MNPT. Effective filtration area: 700 cm²(108¹/₂in.²).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty