Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Swinney Filter Holders, 13 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

4042 4042 4317-N 4317-N
28145-295EA 345.21 USD InStock
CA28144-164 28145-295 CA28145-295 28144-164
Swinney Filter Holders, 13 mm, Cytiva (Formerly Pall Lab)
Filter Holders
The 13 mm Swinney filter holders are economical for small volume (1 to 10 ml) filtration.

  • Available in plastic or stainless steel

They are recommended for filtration of biologicals, ophthalmics, GC and HPLC samples, and lubricants that must be applied dust-free to critical parts. Their luer inlet and outlet fittings provide easy connections.

Materials of Construction Plastic 13 mm Swinney:
- Inlet/Outlet Housing and Support Screen: Celcons (acetal copolymer)
- Seal Washer: PTFE

Materials of Construction for Stainless Steel 13 mm Swinney:
- Body: Type 304 stainless steel
- Gaskets: PTFE
- Support Screen: Photoetched type 304 stainless steel

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty