Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Analyslide®
7231 7231
28145-473PK 125.74 USD InStock
28145-473 CA28145-473
Analyslide® Petri Dish, Cytiva (Formerly Pall Lab)
Culture Dishes Petri Dishes
The Analyslide® Petri dish has a rectangular base and a circular chamber that accepts a 47 mm membrane filter. The sample is protected during microscopic examination, and membrane filters can be stored and protected for reference.
  • Rectangular base fits most microscope stages
  • Inner cover ring for secure closure
  • Clear lid does not interfere with viewing sample
  • Frosted area on base permits identification of the sample with marker
  • Convenient box simplifies collection and storage of samples
Base and lid are constructed from PS.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty