Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pleated Capsule Filters, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

12117 12116 12117 12116
28145-780EA 232.75 USD InStock
28145-780 28145-757 CA28145-757 CA28145-780
Pleated Capsule Filters, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
Designed both for prefiltration of particulate liquids (such as serum) prior to sterilizing filtration, and removal of debris and particles from liquids not requiring sterilization.

  • Maximum operating temperature: 88 °C (190 °F)
  • Maximum operating pressure: 3.4 bar (50 psi)
  • Polycarbonate housing
  • Vented capsules

Pleated Versapor® supported acrylic copolymer membrane filter in a polycarbonate housing

Both capsules are vented and have 1.27 cm (¹/₂") hose barb inlet/outlet connections. Overall length: 21.7 cm (8¹/₂"). The presterilized filter has a hold-up volume of 50 ml, and the nonsterile filter has a hold-up volume of 42 ml.

Packaging: Individually packed, ready-to-use, and disposable.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty