Would you like to log in again?
Supor® Membrane Disc Filters
Supplier: Cytiva
Supor®
66550 66554 66551 66556 66234 60310 61854 60309 60173 60170 60305 60177 66553 60309 66234 60175 60301 66555 60301 60174 60114 66556 60109 66553 60206 60179 66551 60311 60170 66549 60043 60206 61854 60311 60334 60116 60174 66554 60305 66555 60173 60298 60300 60172 60334 66550 60114 60109 60177 60298 66549 60175 60172 60110 60312 60307 65472 60112 65472 66552 66552 60307 60116 60110 60179 60312 60043 60112 60300
28100-004PK 453.15 USD InStock
CA28100-014 28100-004 CA28100-012 28100-002 28100-008 28147-581 28100-006 CA28147-708 76487-590 CA28147-435 28147-469 28147-945 CA28147-956 CA28147-515 76487-592 CA87003-818 CA28147-990 28147-661 28147-980 28147-741 CA28100-010 CA28147-672 28100-000 28147-468 28100-014 28147-708 CA28147-719 28147-956 28147-515 CA76487-592 CA28147-526 87003-818 28147-990 28147-672 CA28147-640 28100-012 28147-435 28100-010 CA87000-002 28146-993 28147-719 28146-992 28146-991 28146-990 CA28147-016 28147-526 CA28147-978 CA28147-617 CA28147-979 28147-640 CA28100-002 CA28100-000 CA28100-006 CA28100-004 CA28146-990 CA28100-008 CA28146-993 CA28146-992 CA28147-468 28147-978 CA28147-469 CA28147-945 28147-617 28147-979 CA28147-581 CA28147-661 CA28147-980 CA28147-741 28147-016
Supor® Membrane Disc Filters
Filters Membrane Filters
Supor® hydrophilic polyethersulfone membranes offer higher fluid flow rates, lower protein binding, lower extractables, and greater strength than other cellulosic (acetate, nitrate) membranes.
  • Hydrophilic
  • Higher flow rate
  • Stronger than cellulose
Provide some of the highest water flow rates and fluid filtration capacities of any membrane filter. These membranes are non sterile, except for 28147-979, which is gamma-irradiated.
The 142 and 293 mm discs feature printed tab for instant recognition of pore size and lot number.
Several of these membranes are offered as recommended replacements for the discontinued HT Tuffyrn membranes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty