Would you like to log in again?
Whatman™ Membrane-Butler, Manual, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10477100 10477100
28151-134EA 1589.35 USD InStock
28151-134 CA28151-134
Whatman™ Membrane-Butler, Manual, Whatman products (Cytiva)
Membrane Dispensers
Manual butler for dispensing filtration membranes.
  • High reliability
  • Cross contamination risks are minimized
  • Membrane dispensed rapidly
  • Highly suitable for use on sterile benches
  • Compact dimensions for portable use
The Membrane-Butler dispenses individual sterile MicroPlus and ME membrane filters (type 'STL'). The sterile packaging is inserted into the roller system and the system is ready to be used.
Membrane filters for microbiological checks must be handled carefully to ensure that they remain sterile and that quantitative results are being obtained.
The Membrane-Butler offers optimal handling for all MicroPlus and ME membrane filters with the type name ‘STL’. The dispenser box is placed in the Membrane-Butler, the sterile packaging is inserted into the roller system, and the system is ready. With each turn (Manual Butler), a membrane filter is ejected from its sterile packing and can be removed easily with forceps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty