Would you like to log in again?
Whatman™ Polyester Drain Discs, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
230600 230800 231100 230300 230600 231100 230500 230500 230300 230800
15000-602PK 79.6 USD InStock
CA28156-243 CA11012-804 15000-602 CA11013-696 CA11013-694 10035-600 10035-602 CA15000-602 10035-598 28156-243
Whatman™ Polyester Drain Discs, Whatman products (Cytiva)
Filters Filter Discs
The polyester drain disc is binder free and has a thickness of 100 µm.
  • Flat surface to eliminate filter tearing or rupturing
  • Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty