Would you like to log in again?
Whatman™ Nylon Membrane Circles, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
7402-002 7404-002 7404-001 7402-001 7402-002 7402-001 7404-004 7408-004 7404-004 7404-009 7402-009 7410-004 7410-004 7404-001 7402-004
28159-723PK 198.55 USD InStock
CA28159-721 CA28159-743 28159-723 28159-746 28159-768 28159-721 28159-743 28159-744 CA28159-720 28159-728 CA28159-746 28159-720 28159-751 10035-550 CA76540-416
Whatman™ Nylon Membrane Circles, Whatman products (Cytiva)
Filters Membrane Filters
High-quality nylon membranes are suitable for filtering aqueous solutions and most organic solvents. The membranes are suitable for use with a wide range of biological preparations and can be used where other membranes are unsuitable or difficult to use.
  • Flexible, durable and tear resistant
  • Can be autoclaved at 135 °C
Nylon membranes are hydrophilic, eliminating the need for wetting agents that could be extracted when filtering aqueous solutions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty