Would you like to log in again?
Activity Wheel for Rodents, Tecniplast®
Supplier: Tecniplast
2154F0105 1284L0105
10709-996EA 1514 USD InStock
10709-996 28196-272
Activity Wheel for Rodents, Tecniplast®
Animal Supplies Animal Cages
Stainless steel construction with low-friction bushings for long life and quiet, smooth action. Stainless steel lid for secure animal containment.
  • Autoclavable
  • Activity wheel allows easy, convenient measurement of lab rodents’ physical activity in response to chemical or environmental stimuli
  • Larger size wheel assembly, for use with size F plastic cage system, accommodates laboratory rats and hamsters
  • Smaller size wheel assembly, for use with size L plastic cage system, accommodates laboratory mice
Ideal for easy monitoring of rodents’ activity levels. Especially useful for research involving circadian rhythm studies, neuroscience, and pharmaceutical testing.
Ordering information: Activity wheel includes wheel, wheel hub and support, sheet lid for secure containment, and wire lid with water bottle support and food hopper. Magnetic switch is sold separately. For plastic cages, see 10712-074 series. Water bottle, stainless steel support rack, and tray (shown in photo) are sold separately. Overall dimensions are for fully assembled cage and wheel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty