Would you like to log in again?
Nalgene® L900 Liquid Detergent, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
900-4000
28196-320CS 397.96 USD InStock
28196-320
Nalgene® L900 Liquid Detergent, Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
Nalgene L900 Liquid Detergent is formulated to thoroughly clean all plastic labware, especially polycarbonate
The detergent will not cause crazing, stress cracking or clouding when used as directed. This liquid detergent are for hand or machine use, it is biodegradable, not toxic, and low foaming.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty