Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Supplier: Thermo Scientific
122-0020 122-0045 121-0045 524-0020 121-0020 124-0045
28199-032CS 898.62 USD InStock
28199-032 28199-034 28199-030 28199-036 73520-980 28199-048
Nalgene® Filter Units, 115 ml, Sterile, Thermo Scientific
Filters Bottle Top Filters
Complete filter units.
  • Choose from CN, aPES, PES, or SFCA membranes
  • Use 0.2 µm pore size for sterile filtration and 0.45 µm pore size for clarification and particle removal
Note: For research use only. Not for in vitro diagnosis or parenterals.
Presterilized, individually packaged one-piece vacuum filter units with 115 ml capacity are ideal for easy, economical filtration of 50 to 115 ml of aqueous fluids. Upper and lower chambers with graduations do not separate; simply remove vacuum hose after filtration and pour filtered media out. Catalog number, membrane, pore size, lot number and expiration date are printed on upper chamber of each filter unit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty