Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® MF76™
450-0020 125-0080 125-0045 126-0045 127-0045 126-0020 450-0080 126-0080 125-0020 450-0045 127-0080 127-0020
28199-098CS 178.55 USD InStock
28199-098 28199-076 28199-155 28199-144 28199-177 28199-280 28199-101 28199-123 28199-112 28199-134 28199-279 28199-268
Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Filters Bottle Top Filters
For easy, fast, sterile filtration of 150 to 1000 ml volumes.
  • Instantly ready for biological filtration, cold sterilization, or cleanup of particulated solutions or buffers
  • Components separate easily for pouring
  • No need to transfer sterile filtrate to another container
  • Graduations on uppers and receiver
  • Non-toxic, triton-free, preassembled and radiation sterilized
Nalgene 1¹/₂-turn threaded screw closure guarantees leakproof seal and eliminates pH shift in receiver bottles. Receiver bottles can be used to store filtrate. Filter cup is printed with catalog number, lot number, membrane type, pore size, and expiration date.
Ordering information: Units with 500 and 1000 ml units include 12 glass fiber prefilters.
Caution: For laboratory use only. Not for in vitro diagnosis or parenterals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty