Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® MF75™
290-3345 291-3320 292-3320 292-4520 291-4545 291-4520 291-3345 290-3320 290-4545 290-4520
28199-296CS 137.69 USD InStock
28199-296 28199-298 28199-307 28199-309 28199-303 28199-317 28199-305 28199-300 28199-302 28199-312
Nalgene® Rapid-Flow™ Bottle-Top Vacuum Filters, Surfactant-Free Cellulose Acetate, Sterile, Thermo Scientific
Filters Bottle Top Filters
SFCA (surfactant-free cellulose acetate) membrane filters are much cleaner than standard cellulose acetate and are designed for vacuum filtration with glass media bottles.
  • Certified radiation-sterilized, non-pyrogenic, and non-cytotoxic
  • Quick-disconnect sidearm accepts ¹/₄ to ³/₈" tubing
  • Use the 0.2 µm size for sterilization of aqueous solutions and culture media
Polystyrene bottle-top design features molded graduations and a support plate that minimizes foaming and eliminates the need for a separate filter pad. Filter unit screws directly onto media bottles. Yellow collar filter cup is labeled with lot number, catalog number, pore size, and expiration date.
Select the 0.45 µm size for particle analysis, scintillation counting, prefiltration of aqueous solutions, and general cytology.
Caution: Use only with sterile bottles designed for vacuum use. Always use safety shield during vacuum procedures. For laboratory use only, not for in-vitro diagnosis or parenterals.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty