Would you like to log in again?
Nalgene® Vacuum Manifold, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS0345-0001 DS0315-0047
28198-910EA 2869.32 USD InStock
28198-910 28199-420
Nalgene® Vacuum Manifold, Thermo Scientific
Manifolds
Designed with three vacuum outlets, each with leakproof, two-way valve and vent port
For simultaneous filtrations with increased filtrate volume in particulate and microbial analysis, use filter funnel with clamp. May also be used with Nalgene® analytical filter funnels or filter holders with funnel.
Each outlet securely holds a No. 8 stopper and the stem of a filter funnel without interference. Outlets are spaced for easy manipulation of individual funnels. Teflon® resin stopcock on each valve shuts off flow when fewer than three outlets are used.
Ordering information: Filter funnel with clamp is packaged as a case of 1.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty