Would you like to log in again?
SAV-IT™ Closures, Porex Bio Products Group
Supplier: Porex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SAV-IT™
A21080 A21072 A21077 A21068 A21075 A21069 A21066 A21074 A21079 A21073 A21065 A21078 A21076 A21070 A21071
28295-458CS 1560.01 USD InStock
28295-458 28295-456 28296-618 28296-606 28296-608 28296-610 28296-620 28296-612 28296-622 28296-614 28296-602 28296-624 28296-616 28296-626 28296-604
SAV-IT™ Closures, Porex Bio Products Group
Closures Test/Sample Tube Closures
Pliable closures for use with evacuated blood collection tubes and glass and plastic test tubes
Seals inside test tube or over tube rim. The 12mm and 13mm closures seal over the rim of either 12mm or 13mm tubes, and when reversed seal inside 10mm tubes. The 16mm closures seal over the rim of 16mm tubes, and when reversed seal inside 12mm and 13mm tubes.
They protect from contamination and prevent evaporation. Closure will not crack or pop off.
Ordering information: Supplied 1000 closures per pack, 10 packs per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty