Would you like to log in again?
FILTER SAMPLER® Blood Serum Filters, Porex
Supplier: Porex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FILTER SAMPLER®
6202 6259 6258 6236 6226 6414 6251 6410 6241 6211 6214 6216 40475
89136-814CS 2653.61 USD InStock
89136-814 28295-612 28295-556 28295-534 28295-501 28295-617 28295-537 28295-658 28295-603 28295-614 28295-526 28295-532 28295-530
FILTER SAMPLER® Blood Serum Filters, Porex
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Self-contained, disposable devices for filtering fibrin and separating serum and plasma from centrifuged whole blood specimens contained in standard 16, 13, or 10.25mm I.D. evacuated blood collection tubes. Technique reduces second centrifugations, tube labeling, and sample preparation time. With collection tube in vertical position, insert stopper end of the filter sampler at a 45° angle and push gently down to just above clot. Filtered serum or plasma will rise into the barrel for analysis.
Tubes come in standard, IB™ (Ion Barrier), and Instrument Specific™ models. Use the standard models when the filtered serum and plasma can be immediately poured off for analysis. The IB™ models help prevent ionic contamination and pH changes and contain a leak-resistant, one-way valve to permit storage of capped specimens in primary collection tube for up to seven days. A physical gap to be created between the serum/plasma and clot, which prevents backflow into the primary collection tube. Instrument Specific™ models permit direct sampling from primary collection tubes and help eliminate specimen relabeling and add-on products as sample cups and tubes. The standard models can be used with glass collection tubes while the IB™ and Instrument Specific™ models can be used with glass or plastic tubes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty