Would you like to log in again?
VWR® Qualitative Filter Paper, Grade 413
Supplier: VWR
VWR®
28310-140 28310-048 28310-015 28310-161 28311-029 28310-106 28310-172 28310-106 28310-081 28310-208 28310-140 28310-015 28309-967 28297-082 28310-081 28309-989 28309-967 28310-048 28310-026 28311-029 28310-208 28309-989 28297-082 28310-026 28310-128 28310-161 28310-128
28310-015PK 13.11 USD InStock
CA28310-048 CA28310-026 28310-015 CA28310-106 CA28310-128 CA28310-208 28297-082 28311-029 CA28310-161 CA28310-140 CA28309-989 CA28309-967 CA28310-081 28310-172 CA28310-015 28310-106 28310-128 28310-048 28310-026 28310-208 CA28297-082 28310-140 CA28311-029 28310-081 28309-967 28309-989 28310-161
VWR® Qualitative Filter Paper, Grade 413
Filters Filter Paper Qualitative Filter Paper
Suitable for filtration of aluminum phosphate, barium carbonate, lead chromate, and more.
  • For routine lab work
Suitable for student requirements in general chemistry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty