Would you like to log in again?
Whatman™ Grade 113V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1213-185 1213-320 1213-185 1213-125 1213-240 1213-270 1213-500 1213-270 1213-150 1213-125 1213-320 1213-240 1213-150 1213-500
28297-278PK 355.62 USD InStock
28297-278 CA28297-276 CA28297-278 CA28467-044 CA28297-270 CA28297-272 CA28297-274 28467-044 28297-276 28297-274 CA10147-914 28297-272 28297-270 10147-914
Whatman™ Grade 113V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Cytiva's Whatman Grade 113V creped filter paper, for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
  • Extremely fast filtration (Herzberg 28s)
  • Wet-strengthened with chemically stable resin
  • Retention of coarse particles: nominal particle retention rating of 30 µm
  • Creped filter paper supports high loading capacity
  • Fluted (pre-pleated) for easy use in applications involving a conical filter funnel
Grade 113V is strengthened with chemically stable resin. However, these paper filters do contain nitrogen and should therefore not be used in Kjeldahl estimations or other nitrogen-sensitive procedures.
Creped filter paper, which has a rough surface, is designed to increase loading capacity. Fluted (pre-pleated) paper filters eliminate the need to quadrant-fold circles in repetitive or multiple analyses
Whatman Grade 113V paper filters from Cytiva's business are further strengthened with chemically stable resin that increases wet-strength without introducing significant impurities to the filtrate. Strengthening the paper expands the range of sample types that can be separated in a conical filter funnel.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty