Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 540, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™
1540-090 1540-185 1540-150 1540-321 1540-240 1540-150 1540-321 1540-125 1540-324 1540-320 1540-055 1540-324 1540-185 1540-125 1540-090 1540-110 1540-042 1540-110 1540-055 1540-240 1540-042 1540-320
10147-986PK 44.7 USD InStock
CA70240-064 CA10147-890 10147-986 28297-368 28495-023 28495-045 CA28297-376 10147-890 CA28297-368 70240-064 CA28297-370 CA28297-372 10035-282 CA28297-374 28297-376 CA28495-023 CA28495-045 28297-374 CA10147-986 CA11013-738 28297-372 28297-370
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 540, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 540 is a high-purity, acid-treated cellulose lab filter paper. Mechanically strong Grade 540 is well suited for quantitative or gravimetric analysis of strongly acidic or alkaline samples.

  • Acid-treated for high wet-strength
  • Extremely low ash content (0.006% ash maximum)
  • Nominal particle retention of 8 µm
  • Medium flow rate (Herzberg filtration speed 200 s)
  • High chemical resistance to strong acid and alkali suspensions

Cytiva's business offers hardened ashless filter paper for a variety of precipitate sizes. Along with general filtration Grade 540, the range includes Grade 542 for retention of fine precipitates and Grade 541 for fast filtration. All three grades are designed for use in gravimetric analysis.

Cytiva's hardened, ashless Grade 540 lab filter paper is frequently used as a gravimetric filter during analysis of metals in acidic or alkaline solutions and in the collection of hydroxides. The acid-treated surface prevents tearing during filtration and allows the user to scrape precipitates from the paper.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty