Would you like to log in again?
Whatman™ High-Purity Glass Soxhlet Extraction Thimbles, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
2814-199 2814-432 2814-432 2814-533 2814-300 2814-259 2814-259 2814-300 2814-533 2814-199
27732-185PK 327.09 USD InStock
27732-185 CA27732-185 CA28297-440 10035-424 28297-440 CA11012-884 CA27732-190 27732-190 CA11012-886 10035-422
Whatman™ High-Purity Glass Soxhlet Extraction Thimbles, Whatman products (Cytiva)
Extractors Extraction Thimbles
Cytiva's Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
  • Resistant to temperatures up to 500° C
  • Binder-free, no chemical additives
  • Designed to fit most commercially available Soxhlet extractors
  • Effective when used with solvents incompatible with cellulose materials
Glass Soxhlet extraction thimble applications:
Analysis of hot and acidic gases during emission control and smoke stack gas monitoring
Analysis of pesticide residues using Soxhlet extraction
Determination of oil/fat content of foods
Analysis of oil and grease in solid wastes
Whatman brand high-purity glass thimbles are available in a range of sizes and wall thicknesses for optimal use in a variety of air sampling and Soxhlet extraction units.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty