Would you like to log in again?
VWR® Quantitative Filter Paper, Grade 494
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
28297-930 28307-134
28297-930PK 51.56 USD InStock
28297-930 28307-134
VWR® Quantitative Filter Paper, Grade 494
Filters Filter Paper Quantitative Filter Paper
Useful for very fine precipitates such as barium and lead sulfates, stannic and nickel sulfides, and calcium hydroxide.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty