Would you like to log in again?
VWR® Quantitative Filter Paper, Grade 454
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
28306-110 28306-175 28306-175 28306-153 28306-131 28306-095 28306-051
28306-175PK 89.12 USD InStock
28306-175 28306-153 28306-131 28306-110 CA28306-175 28306-095 28306-051
VWR® Quantitative Filter Paper, Grade 454
Filters Filter Paper Quantitative Filter Paper
Use for retention of zirconium hydroxide, aluminum hydroxide, cobalt sulfide, and other coarse or gelatinous precipitates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty