Would you like to log in again?
VWR® Quantitative Filter Paper, Grade 474
Supplier: VWR
VWR®
28306-368 28306-380 28306-346 28306-380 28306-404 28306-346 28306-404 28306-368
28306-380PK 49.5 USD InStock
CA28306-404 28306-380 CA28306-380 CA28306-346 CA28306-368 28306-404 28306-368 28306-346
VWR® Quantitative Filter Paper, Grade 474
Filters Filter Paper Quantitative Filter Paper
Uses include filtration of aluminum phosphate, zinc ammonium phosphate, and silver chromate.
For general quantitative analyses.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty