Would you like to log in again?
VWR® Qualitative Crepe Filter Paper, Grade 417
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
28297-868 28313-068 28297-868 28313-240 28313-104 28313-160 28313-206 28313-080 28313-148 28313-046 28313-206 28313-080 28313-181 28313-126 28313-160 28313-148 28313-057 28313-068 28313-181 28313-104 28313-240 28313-046 28313-057 28313-126
28297-868PK 12.09 USD InStock
28297-868 CA28297-868 CA28313-206 28313-206 CA28313-104 CA28313-148 CA28313-126 CA28313-068 CA28313-046 28313-148 CA28313-057 28313-104 28313-126 28313-068 28313-046 28313-057 CA28313-240 28313-240 CA28313-160 CA28313-181 28313-181 28313-160 CA28313-080 28313-080
VWR® Qualitative Crepe Filter Paper, Grade 417
Filters Filter Paper Qualitative Filter Paper
Good for rapid filtrations where fine particle retention is not required.
  • Coarse porosity
  • Fast flow rate
  • Assorted sizes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty