Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Grade Shark Skin Filter Papers for Technical Use, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™ Sharkskin™ Sharkskin™
10347509 10347525 10347519 10537138 10347530 10347521 10347510 10347530 10347522 10347577 10347511 10537138 10347523 10347577 10347513 10347521 10347511 10347519 10347509 10347512 10347522 10347510 10538877
28314-050PK 68.5 USD InStock
28314-050 28314-061 28314-083 74300-028 CA74300-028 28314-108 28314-028 CA28314-028 CA89125-938 89125-938 28314-184 74330-294 28314-141 28314-152 28314-163 CA28314-061 CA28314-050 CA28314-163 28314-186 CA28314-152 CA28314-141 28314-220 CA28314-184
Whatman™ Grade Shark Skin Filter Papers for Technical Use, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Whatman Grade Shark Skin filter papers are a creped filter paper that is commonly used to remove impurities in a variety of applications.

  • Very thin with high wet strength: Enables handling when wet; allows use under pressure; provides durability and structural support in aqueous filtration
  • Creped filter paper: Has enlarged surface area that reduces duration of filtration and allows more stretch and flexibility
  • Durable: Resistant to weak acids and bases

Whatman Grade Shark Skin Filter Papers (particle retention 8-12 µm) are a medium/slow filter paper made from super-refined cellulose. They are available in a variety of sizes, in both circles and sheets.

Applications include:
- Purification of edible oils, beverages, and cocoa butter for food and cosmetic uses.
- Removal of suspended solids for purification of electroplating baths
Protective paper for filter press cloths
- Reducing impurities lowers the amounts of reactants and increases the efficiency of the oil, which minimizes production costs and helps to ensure regulatory compliance.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty