Would you like to log in again?
Whatman™ Grade 202 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
5202-185 5202-110 5202-250 5202-200 5202-320 5202-125 5202-250 5202-320 5202-090 5202-090 5202-200 5202-125 5202-240 5202-150 5202-240 5202-400 5202-150 5202-090 5202-330 5202-330 5202-185 5202-110 5202-150 5202-110 5202-125 5202-400
28317-320PK 66.76 USD InStock
28317-320 470105-676 CA28317-230 470205-548 28317-263 28317-241 470105-678 28317-284 28500-050 CA28317-309 CA11008-524 CA28297-572 CA28500-047 28500-038 10147-948 CA28317-241 CA28317-263 CA28317-284 28317-230 CA28500-050 CA28317-320 28317-309 CA28500-038 28297-572 470205-550 28500-047
Whatman™ Grade 202 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A general qualitative filter with creped surface. Faster than grade 201 and retains large crystalline coarse particles. Widely used in industrial laboratories and students laboratories for routine filtration.
  • Filters coarse particles
  • Medium filtration speed
  • Creped for increased efficiency
The filter is wet-strengthened with a small amount of chemically stable resin to extend the life of the filter and improve handling. Normal qualitative applications will not introduce any significant impurities into the filtrate. However it is not recommended for Kjeldahl nitrogen analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty