Would you like to log in again?
Whatman™ Lids, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ UniSeal™ UniSeal™
7704-1001 7704-1001
28317-488PK 213.57 USD InStock
CA28317-488 28317-488
Whatman™ Lids, Whatman products (Cytiva)
Lids Microplate Lids
Universal lids suitable for robotic handling. Can be used as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty