Would you like to log in again?
VWR® Qualitative Crepe Filter Paper, Grade 415
Supplier: VWR
VWR®
28320-019 28320-100 28320-223 28320-020 28320-187 28320-121 28320-143 28320-187 28297-846 28320-020 28320-201 28297-846 28320-019 28320-085 28320-041 28320-085 28320-100 28320-143 28320-223 28320-121 28320-201 28320-041
28297-846PK 114.67 USD InStock
28297-846 CA28297-846 28320-085 28320-041 CA28320-085 CA28320-041 28320-020 28320-100 28320-121 28320-187 CA28320-020 28320-143 CA28320-019 CA28320-100 28320-223 28320-201 CA28320-187 CA28320-143 CA28320-121 CA28320-201 CA28320-223 28320-019
VWR® Qualitative Crepe Filter Paper, Grade 415
Filters Filter Paper Qualitative Filter Paper
Good for coarse precipitates such as aluminum hydroxide, cobalt carbonate, mercury iodide, magnesium hydroxide, and more.
  • Coarse porosity
  • Fast flow rate
  • Assorted sizes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty