Would you like to log in again?
VWR® Qualitative Filter Paper, Grade 410
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
28321-113 28297-826 28321-077 28321-113 28321-135 28321-135 28321-077 28297-826
28321-113PK 23.64 USD InStock
CA28321-113 CA28321-135 CA28297-826 28321-113 28321-135 CA28321-077 28321-077 28297-826
VWR® Qualitative Filter Paper, Grade 410
Filters Filter Paper Qualitative Filter Paper
Grade 410 filter paper is useful in analytical operations involving filtration of cuprous oxide, lead sulfate, barium sulfate, manganese carbonate, and more.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty