Would you like to log in again?
VWR® Qualitative Crepe Fluted Circles, Folded, Grade 315
Supplier: VWR
VWR®
28331-048 28331-128 28331-026 28331-106 28331-106 28331-048 28331-081 28331-060 28331-081 28331-128 28331-060 28331-026
28331-060PK 138.57 USD InStock
28331-060 CA28331-081 CA28331-060 CA28331-048 CA28331-026 28331-128 28331-106 CA28331-106 28331-081 CA28331-128 28331-026 28331-048
VWR® Qualitative Crepe Fluted Circles, Folded, Grade 315
Filters Filter Paper Qualitative Filter Paper
Crepe fluted circles are ideal for gelatinous and coarse precipitates.
  • Coarse porosity
  • Fast flow rate
  • Assorted sizes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty