Would you like to log in again?
VWR® Qualitative Fluted Circles, Folded, Grade 313
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
28333-043 28333-087 28333-043 28333-065 28333-021 28333-087 28333-021 28333-123 28333-065 28333-123 28333-101
28333-087PK 135.22 USD InStock
28333-087 28333-043 28333-021 28333-065 CA28333-043 CA28333-065 CA28333-021 CA28333-087 28333-123 28333-101 CA28333-123
VWR® Qualitative Fluted Circles, Folded, Grade 313
Filters Filter Paper Qualitative Filter Paper
Used for drugs, pharmaceuticals, liquors, tinctures, chromatography, and protein-free filtrations in the clinical laboratory.
  • Medium fine porosity
  • Medium flow rate
  • Assorted sizes
Fluted circles are available in a variety of sizes and boast a medium flow rate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty