Would you like to log in again?
VWR® Glass Fiber Filters
Supplier: VWR
VWR®
28333-137 28297-500 28297-982 28297-974 28297-978 28333-129 28297-500 28333-133 28333-127 28297-508 28297-506 28333-131 28333-139 28297-504 28333-131 28297-510 28297-976 28333-125 28297-976 28333-135 28333-129 28333-133 28297-508 28297-506 28297-982 28333-127 28297-289 28333-135 28297-984 28333-125 28297-504 28297-978 28297-984 28333-139 28297-510 28297-974 28297-289 28333-137
28297-504PK 99.46 USD InStock
28297-504 CA28297-978 28297-500 28297-984 28297-982 28297-289 CA28297-510 CA28297-974 28297-508 CA28297-976 28297-506 28333-129 CA28333-139 CA28333-131 CA28333-133 28333-125 CA28333-135 28333-127 CA28333-137 28297-978 CA28297-504 CA28297-506 28297-976 CA28297-508 28297-974 28297-510 28333-131 CA28297-982 CA28297-289 CA28297-500 CA28297-984 CA28333-127 CA28333-129 28333-133 28333-135 28333-137 CA28333-125 28333-139
VWR® Glass Fiber Filters
Filters Glass Fiber Filters
Binder-free VWR® glass fiber filters feature fast flow rates, high-loading capacity, wide thermal tolerance, and excellent precipitate retention.
  • Fast flow rates
  • High-loading capacity
  • Wide thermal tolerance
  • Excellent precipitate retention
Available in three grades: Grade 691 is an ideal filter for radioimmunoassay of weak beta emitters by scintillation counting, and for filtration of precipitated proteins, cells, and cell debris. Also used for gravimetric determination of airborne particulates. Grade 693 is recommended for use in dissolved and suspended solids testing in sanitary water analysis procedures. Grade 696 is used for monitoring specific pollutants (e.g., Hg in water), and in marine chemistry to filter particulate components in seawater. Also used in RIA procedures and for harvesting lymphocytes during incorporation trials.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty