Would you like to log in again?
FILTER PAPER GRADE 601
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
GR601 Filter Paper^ Use in Electrophoresis/Blotting, and Qualitative and Seed Germination. Slow to Medium absorbance rate. Weight 88g/m2, Thickness 0.19mm, Rapidity 55ml/min, Capillary Rise 21mm, 100% Cotton fiber, 3/4 inch wide roll, 1000 feet.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER PAPER GRADE 601 28340-012 601-34 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty