Would you like to log in again?
FILTER PAPER 1/2IN DIA PK1000
Supplier: Cytiva
Grade 740-E Special-Purpose Filter Paper^ 1.27cm diameter. Thick, absorbent cotton linter for use as an antibiotic assay test disc. Specified in standard tests for antibiotic susceptibility as well as antibiotic levels in milk. Coarse porosity. Fast flow rate. 20-30um particle retention.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER PAPER 1/2IN DIA PK1000 28446-006 10328170 Pack of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty