Would you like to log in again?
Whatman™ Grade 1 Qualitative Filter Paper, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1001-030 1001-045 1001-125 1001-240 1001-917 1001-090 1001-385 1001-824 1001-055 1001-320 1001-813 1001-070 1001-325 1001-6508 1001-085 1001-500 1001-030 1001-400 1001-329 1001-020 1001-734 1001-047 1001-385 1001-320 1001-025 1001-047 1001-085 1001-0155 1001-270 1001-042 1001-020 1001-032 1001-931 1001-032 1001-329 1001-110 1001-110 1001-824 1001-045 1001-055 1001-070 1001-090 1001-025 1001-125 1001-929 1001-240 1001-185 1001-500 1001-0155 1001-325 1001-150 1001-400 1001-918 1001-270 1001-150 1001-813 1001-932 1001-185 1001-917 1001-932 1001-931 1001-6508 1001-042 1001-734 1001-929 1001-918
28450-150PK 40.61 USD InStock
28450-150 CA11012-592 CA11012-594 CA28450-160 28450-070 CA28451-006 CA28450-081 CA11012-596 76423-768 11008-512 10035-810 CA28450-207 CA28450-128 10035-218 10035-812 CA28450-048 28450-235 CA28450-331 10035-818 89233-648 28297-216 10035-814 10035-816 28450-081 CA76423-768 28450-160 28451-006 CA11008-512 10035-220 CA21426-402 28450-207 10035-222 CA28450-015 28450-048 CA89233-648 28450-128 10035-224 CA70240-010 28450-331 CA28450-021 CA11012-890 CA11012-892 CA28450-182 CA11012-896 CA11012-810 CA28297-216 CA70240-096 CA11012-812 CA11012-808 CA28450-229 21426-402 CA28450-106 CA70240-008 CA28450-026 28450-015 CA28450-150 28450-021 CA28450-070 28450-182 70240-096 70240-010 28450-229 70240-008 CA28450-235 28450-026 28450-106
Whatman™ Grade 1 Qualitative Filter Paper, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Whatman Grade 1 Qualitative Filter Papers are the most widely used filter paper for routine laboratory applications, qualitative analytical separations, and clarifying liquids. They are the standard grade cellulose filter papers for medium flow rates.
  • Qualitative analytical separations for precipitates such as lead sulfate, calcium oxalate (hot), and calcium carbonate
  • Agricultural soil analysis and seed testing procedures
  • Industrial food processing techniques to extract liquid or separate solid foodstuffs from associated liquid
  • Air pollution monitoring by collecting atmospheric dust from airflow and photometrically measuring the stain-intensity
  • Gas detection by quantitative chromogenesis with optical reflectance
Grade 1 Qualitative Filter Papers (11 µm particle retention) are available in a variety of sizes and formats including circles, sheets and prepleated filters. They are also available in FilterCup format, a convenient, 250 ml capacity disposable 70 mm filter funnel molded from polypropylene with an integral, heat-bonded filter.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty