Would you like to log in again?
Whatman™ Membrane Filters, PTFE, WTP Range, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
7590-004 7582-004 7590-002 7590-004 7582-004 7582-002 7582-002 7585-004 7585-004 7590-002
28454-430BX 694.45 USD InStock
CA11008-532 CA28454-430 28454-430 CA28454-450 CA28454-470 28454-450 28454-470 CA28454-418 28454-418 11008-532
Whatman™ Membrane Filters, PTFE, WTP Range, Whatman products (Cytiva)
Filters Membrane Filters
Whatman PTFE membranes from Cytiva are strong hydrophobic membrane filters resistant to the corrosive solvents and aggressive media used in gas sterilization and liquid clarification.
  • Polypropylene grid support material for increased strength
  • Temperature resistant up to 120 °C
  • Resistant to most aggressive acids, alkalis, and solvents
  • Hydrophobic membrane filters can collect aqueous aerosols
PTFE membranes are especially effective for use with aggressive solvents, liquids, and gases.
PTFE membranes are used for liquid clarification of corrosives, solvents, and aggressive fluids. Effective sample filtration is critical to prevent equipment damage. Cytiva's PTFE hydrophobic membrane filters are also suitable for air and gas sterilization because of their ability to capture aqueous aerosols.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty