Would you like to log in again?
Whatman™ Grade 3 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1003-320 1003-323 1003-070 1003-185 1003-917 1003-323 1003-070 1003-240 1003-185 1003-110 1003-090 1003-917 1003-150 1003-055 1003-090 1003-240 1003-055 1003-320 1003-110 1003-125 1003-150 1003-125
28297-229PK 825.97 USD InStock
CA28297-229 28297-229 70240-016 CA70240-016 CA89013-946 CA28456-043 CA28456-065 28456-022 CA28456-087 28456-043 28456-087 28456-065 CA28456-167 CA28456-145 CA28456-123 CA28456-101 28456-145 28456-167 28456-101 28456-123 CA28456-022 89013-946
Whatman™ Grade 3 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
  • Thickness of 6 µm
Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Double the thickness of Grade 1 with still finer particle retention and excellent loading capacity; more precipitate can be held without clogging. The extra thickness gives increased wet strength and makes this grade highly suitable for use in Büchner funnels.
The high absorbency is particularly valuable when the paper is used as a sample carrier. This filter is also available in the FilterCup. This is a convenient, disposable 70 mm filter funnel with a 250 ml capacity molded from polypropylene with an integral, heat bonded filter (catalog number 1600-003).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty