Would you like to log in again?
Whatman™ Grade 4 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1004-125 1004-027 1004-648 1004-648 1004-047 1004-110 1004-090 1004-070 1004-150 1004-185 1004-050 1004-917 1004-047 1004-320 1004-090 1004-055 1004-070 1004-930 1004-027 1004-400 1004-270 1004-041 1004-125 1004-055 1004-150 1004-110 1004-917 1004-042 1004-270 1004-050 1004-930 1004-041 1004-400 1004-320 1004-042 1004-185 1004-240 1004-240 1004-150
70240-024PK 144.78 USD InStock
70240-024 10035-824 70240-026 28460-142 28460-164 28460-120 CA28460-030 28460-063 28460-041 CA11012-830 28460-085 CA28460-171 CA28460-110 10035-230 11008-514 CA11012-606 10035-232 CA11012-608 CA11012-828 CA28460-019 10147-900 28460-171 CA70240-024 CA70240-026 28460-110 CA28460-085 CA28460-041 470019-894 CA28460-063 28460-030 CA11012-602 CA28460-120 CA28460-142 CA28460-164 28460-019 CA11008-514 CA10147-900 10035-226 10035-228
Whatman™ Grade 4 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Whatman Grade 4 Qualitative Filter Papers are a standard grade, high flow rate filter paper. They are useful in qualitative analytical separations when fast flow is required and the collection of fine particles is not critical.
  • Excellent retention: Captures coarse particles and gelatinous precipitates such as ferric hydroxide and aluminum hydroxide
  • Fast filtration: Useful for routine clean-up of biological fluids or organic extracts during analysis
These cellulose filters are used in qualitative analytical techniques to determine and identify materials (e.g., in air pollution monitoring).
Grade 4 Qualitative Filter Papers (20 - 25 µm particle retention) are available in a variety of sizes and formats including circles, sheets, and rolls. They are also available as prepleated filters, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty