Would you like to log in again?
Whatman™ Grade 5 Qualitative Filter Papers
Supplier: Whatman products (Cytiva)
Whatman™
1005-320 1005-060 1005-110 1005-110 1005-070 1005-320 1005-125 1005-150 1005-240 1005-090 1005-055 1005-070 1005-325 1005-240 1005-047 1005-325 1005-185 1005-055 1005-042 1005-042 1005-047 1005-125 1005-185 1005-060 1005-090 1005-150
28462-104PK 50.8 USD InStock
CA70240-034 CA76423-832 28462-104 28462-126 28462-148 28462-047 28462-069 28462-025 28462-166 28462-021 CA28462-030 28462-160 28462-080 70240-034 CA28462-104 CA28462-148 CA28462-126 76423-832 CA28462-047 CA28462-025 CA28462-069 CA28462-166 CA28462-021 CA28462-160 28462-030 CA28462-080
Whatman™ Grade 5 Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
A standard grade clarifying filter excellent for cloudy suspensions and for water and soil analysis.
  • Thickness of 2.5 µm
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
The maximum degree of fine particle filtration in the qualitative range. Capable of retaining the fine precipitates encountered in chemical analysis. Slow flow rate. Excellent clarifying filter for cloudy suspensions and for water and soil analysis. Also available prepleated as Grade 5V.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty