Would you like to log in again?
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Supplier: Whatman products (Cytiva)
Whatman™
1541-070 1541-047 1541-400 1541-240 1541-240 1541-185 1541-125 1541-090 1541-270 1541-150 1541-400 1541-917 1541-917 1541-110 1541-042 1541-320 1541-125 1541-055 1541-150 1541-320 1541-042 1541-055 1541-185 1541-070 1541-270 1541-047 1541-110 1541-090
28479-140PK 304.75 USD InStock
CA28479-130 CA28479-151 28479-140 28479-162 28479-209 CA28479-016 28479-107 28479-129 28479-027 28479-005 28479-049 CA28478-997 CA28479-209 28479-082 28479-060 CA28479-162 CA28479-140 28479-151 28479-130 CA28479-129 CA28479-107 CA28479-027 CA28479-005 CA28479-049 28479-016 28478-997 CA28479-082 CA28479-060
Whatman™ Quantitative Filter Papers, Hardened Ashless Grades, Grade 541
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 541 acid hardened cellulose filter paper features high wet-strength, high chemical resistance, and a large nominal particle retention for fast analytical filtration.
  • High wet-strength
  • High chemical resistance to strong acid and alkali suspensions
  • Retention of gelatinous precipitate
  • 22 µm nominal particle retention rating
  • Fast flow rate (Herzberg filtration speed 34 s)
  • Maximum of 0.006% ash concentration
Along with fast Grade 541 filter paper, the hardened, ashless range from Cytiva's business includes general-purpose Grade 540 and Grade 542 for fine particle retention. All three paper grades are appropriate for use in gravimetric analysis with strong acids and alkali samples.
Hardened ashless filter paper is recommended when fast filtration of a gelatinous precipitate for gravimetric analysis is performed on a strongly acidic or alkali solution.
Applications of Grade 541 papers include:
- Use as an animal foodstuff filter for determining fiber content
- Determination of gelatin content in milk and cream
- Quantification of chloride in cement, as well as chloride and phosphorous in coal and coke
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty