Would you like to log in again?
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 42
Supplier: Whatman products (Cytiva)
Whatman™
1442-047 1442-917 1442-6551 1442-110 1442-042 1442-240 1442-125 1442-042 1442-6551 1442-240 1442-320 1442-320 1442-110 1442-090 1442-070 1442-047 1442-055 1442-090 1442-055 1442-070 1442-185 1442-150 1442-150 1442-125 1442-917 1442-185
10035-270PK 127.76 USD InStock
CA11013-634 CA28480-026 CA28480-048 CA11013-752 CA28480-140 CA28480-060 CA28480-081 CA28480-161 CA28480-182 CA28297-312 CA11008-522 10035-270 10035-272 CA28480-106 CA28480-128 28480-026 28480-048 28297-312 28480-182 28480-161 28480-140 28480-081 28480-060 11008-522 28480-106 28480-128
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 42
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 42 quantitative papers have the finest retention of all Whatman ashless filter paper grades. Grade 42 is designed for gravimetric analysis and sample preparation for instrumental analysis.
  • Fine (2.5 µm) nominal particle retention rating, eliminates large particles that might otherwise interfere with sensitive instrumentation
  • Commonly used as a calcium carbonate filter
These filter papers are designed for retention of extremely small particles. Other typical precipitates for gravimetric analysis include barium sulfate and metastannic acid for use in glazes, coatings and pigments. The high purity of these filters also makes them suitable for processes that require ashing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty