Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 44, Whatman Products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™
1444-150 1444-240 1444-185 1444-090 1444-090 1444-125 1444-185 1444-240 1444-110 1444-125 1444-150 1444-070 1444-110 1444-070
28482-021PK 71.83 USD InStock
CA70240-054 28482-021 28482-065 28482-043 CA28482-021 CA28482-065 CA28482-043 28297-334 CA89233-652 89233-652 CA28297-334 28482-070 70240-054 CA28482-070
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 44, Whatman Products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 44 quantitative paper is a thin version of Grade 42 ashless paper. The thinness reduces the overall ash content, making Grade 44 well suited for analyses relying on ash-free dry weight.

  • Accurate ash free dry mass measurements
  • Fine nominal particle retention rating of 3 µm
  • Filtration speed (Herzberg 995 s)
  • Typical thickness of 180 µm

Cytiva's business offers other ashless grades with different particle retention ratings, as well as hardened ashless grades to ensure accurate measurement during analyses of samples that are strongly acidic/alkali.

Fast gravimetric analysis of fine particles. Gravimetric analysis can be time-consuming when filtration of fine particles is required. Because of its reduced thickness, Grade 44 ashless filter paper from Cytiva provides faster filtration of fine particles when compared with thicker papers of similar retention rating, such as Whatman Grade 42 paper.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty