Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 50, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™
1450-185 1450-090 1450-125 1450-320 1450-070 1450-110 1450-055 1450-993 1450-916 1450-150 1450-240 1450-917 1450-320 1450-150 1450-125 1450-055 1450-500 1450-917 1450-070 1450-063 1450-042 1450-185 1450-042 1450-240 1450-090 1450-993 1450-110 1450-916 1450-063 1450-500
70240-144PK 44.22 USD InStock
70240-144 CA11013-358 CA28485-100 CA28485-121 CA28485-165 CA28485-143 CA11013-356 28485-019 76419-604 10035-274 10035-276 CA28485-020 10035-278 CA28485-063 CA28485-041 CA28485-085 CA76419-604 CA70240-144 CA11013-644 CA11013-760 CA28485-019 10035-280 28485-041 28485-063 28485-085 28485-143 28485-121 28485-165 28485-100 28485-020
Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 50, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Whatman Grade 50 quantitative hardened low ash filter paper is highly glazed to enable recrystallization of very fine precipitates using Büchner funnel or Hirsch funnel filtration.

  • Thinner filter design (average thickness 115 µm) than most Büchner filter papers
  • Nominal particle retention of 2.7 µm
  • Slow flow rates (Herzberg filtration speed 2685 s)
  • High purity and low ash content (0.015% ash maximum)
  • Acid-treated for high wet-strength and chemical resistance
  • Hardened to minimize fiber shredding

This hard, very thin, high wet-strength Grade 50 quantitative filter paper withstands wet handling and precipitate removal by scraping. With very minimal fiber shedding, they are an especially good choice for use as carriers in integrated circuits.

Whatman low ash hardened filter paper for quantitative analysis is available in three grades. All three are designed for use in gravity and vacuum suction filtration in Buchner and Hirsch funnels. Grade 50 is also available in Smear Tab format for wipe testing, such as surface testing for radionuclide contamination.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty