Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 52

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™
1452-090 1452-150 1452-125 1452-150 1452-110 1452-125 1452-110 1452-090 1452-240 1452-240
70240-066PK 73 USD InStock
70240-066 CA70240-066 CA28487-080 28487-080 CA28487-105 28297-354 28487-105 CA28297-354 CA28487-047 28487-047
Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 52
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 52 is a general-purpose filter paper for quantitative analysis in Büchner funnel and other vacuum filtrations. The acid-hardened surface and low ash content enable efficient precipitate recovery.

  • Manufactured from high-quality cotton linters and treated to remove trace metals
  • High wet-strength and chemical resistance
  • Nominal particle retention of 7 µm
  • Medium flow rate (Herzberg filtration speed 235 s)
  • High purity (0.015% ash maximum)
  • Compatible with Hirsch or Büchner funnel applications

Cytiva's business offers three grades of hardened low ash filter paper. All three are designed for use in Hirsch and Büchner funnel applications and can be customized for an exact fit.

The quality of analysis resulting from vacuum filtration using a Hirsch or Büchner funnel relies on using a high-quality filter that avoids introducing artifacts into measurement. Cytiva's Grade 52 hardened filter paper is acid-treated to prevent fiber shredding during vacuum filtration or precipitate collection.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty