Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 54

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™
1454-125 1454-055 1454-070 1454-500 1454-110 1454-070 1454-240 1454-150 1454-090 1454-150 1454-185 1454-917 1454-320 1454-090 1454-055 1454-185 1454-917 1454-320 1454-240 1454-125 1454-110 1454-500
70240-070PK 1052.29 USD InStock
70240-070 CA28297-364 CA70240-070 28489-009 CA89008-904 CA28489-009 28489-100 28489-122 28489-144 CA28489-020 28297-364 CA28489-042 CA28489-086 CA28489-064 28489-086 28489-020 28489-064 28489-042 CA28489-144 CA28489-100 CA28489-122 89008-904
Whatman™ Quantitative Filter Papers, Hardened Low Ash Grades, Grade 54
Filters Filter Paper Quantitative Filter Paper
Whatman Grade 54 hardened low ash quantitative filter paper for Büchner funnel and Hirsch funnel filtration enables fast vacuum suction filtration for recrystallization applications.

  • Higher nominal particle retention rating (22 µm) than comparable Buchner funnel filter papers
  • Fast filtration speed (Herzberg filtration speed 39 s)
  • High purity (0.015% ash maximum)
  • Acid-treated for high wet-strength and chemical resistance
  • Hardened to minimize fiber shedding

This hard, smooth, high wet-strength Grade 54 fast filter paper enables collection of coarse and gelatinous precipitates with minimal fiber shredding, even when wet.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty