Would you like to log in again?
Whatman™ EPM 2000 Air Sampling Filter, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1882-047 1882-866 1882-866 1882-047
28495-420PK 939.27 USD InStock
CA28495-330 28495-420 CA28495-420 28495-330
Whatman™ EPM 2000 Air Sampling Filter, Whatman products (Cytiva)
Filters Glass Fiber Filters
Cytiva's Whatman EPM 2000 air filters are used in high-volume PM-10 sampling equipment.
  • Air retention efficiency of 99.95 percent of 0.3 µm size particulates
  • Selected by the U.S. Environmental Protection Agency (EPA) for use in a nationwide network of HiVol air samplers
  • Manufactured according to ISO9001:2008 QMS standards
  • Available in both sheet and circle design
  • Sheets are individually numbered to facilitate identification
  • Binder-free
EPM 2000 from Cytiva's business is available as 100-piece packs of either 47 mm filter paper circles or 8 10" sheets.
Manufactured from 100% pure borosilicate glass, they enable detailed chemical analysis of trace pollutants with minimal interference.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty