Would you like to log in again?
Whatman™ Glass Microfiber Prefilter, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
1842-090 1841-047 1842-090 1841-090 1842-047 1841-047 1841-090 1842-047
28495-612PK 81.97 USD InStock
CA28495-616 28495-612 28495-622 CA28495-618 CA28495-622 28495-618 CA28495-612 28495-616
Whatman™ Glass Microfiber Prefilter, Whatman products (Cytiva)
Filters Glass Fiber Filters
GMF 150 prefilters are multilayer graded density filters for rapid and efficient filtration of samples with high solids content. They are designed with a course top layer (10 µm) and meshed with a finer layer of 1 or 2 µm. Manufactured from 100% borosilicate glass microfibre, the filter is binder free. It is an excellent prefilter for higher particulate loading capacity with faster flow rates.
  • Nearly doubles the volume of sample filtered compared to a single density prefilter
  • Compared to an unprotected membrane, three to seven times greater sample volume can be filtered
  • Higher particulate loading capacity and faster flow rates than uniform prefilters
  • Excellent particle retention (both large and small particles)
  • Extends life of final filter membrane
When a sample is applied, the multigrade glass filter traps larger particles in its pores or on the surface of the coarse layer, medium sized particles in the interface meshing and smaller particles in the interstices of the fine layer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty